4-(Methylsulphonyl)-3-piperazinobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR470065-5G
APOSOR470065-5GEA
1180
EUR
APOSOR470065-5G
4-(Methylsulphonyl)-3-piperazinobenzoic acid
4-(Methylsulphonyl)-3-piperazinobenzoic acid
Formel:
C₁₂H₁₆N₂O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD13191609 CAS-Nummer: 1197193-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...