4-Bromo-2-piperazinobenzoic acid
Lieferant: Apollo Scientific
OR470066-5G
APOSOR470066-5GEA
804
EUR
APOSOR470066-5G
4-Bromo-2-piperazinobenzoic acid
4-Bromo-2-piperazinobenzoic acid
Formel:
C₁₁H₁₃BrN₂O₂ Molecular Weight: 285,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13191612 CAS-Nummer: 1197193-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...