4-Chloro-2-piperazinobenzoic acid
Lieferant: Apollo Scientific
OR470070-5G
APOSOR470070-5GEA
804
EUR
APOSOR470070-5G
4-Chloro-2-piperazinobenzoic acid
4-Chloro-2-piperazinobenzoic acid
Formel:
C₁₁H₁₃ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13191620 CAS-Nummer: 1197193-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...