2-Chloro-4-piperazinobenzoic acid

Lieferant: Apollo Scientific

OR470072-5G
APOSOR470072-5GEA 804 EUR
APOSOR470072-5G
2-Chloro-4-piperazinobenzoic acid
2-Chloro-4-piperazinobenzoic acid
Formel: C₁₁H₁₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12198409
CAS-Nummer: 74803-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR