Methyl 1-Benzyl-2-piperidineacetate

Lieferant: Apollo Scientific

OR470076-5G
APOSOR470076-5GEA 713 EUR
APOSOR470076-5G
Methyl 1-Benzyl-2-piperidineacetate
Methyl 1-Benzyl-2-piperidineacetate
Formel: C₁₅H₂₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13191628
CAS-Nummer: 247259-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR