1-Benzyl-2-piperidineacetic acid

Lieferant: Apollo Scientific

OR470077-5G
APOSOR470077-5GEA 804 EUR
APOSOR470077-5G
1-Benzyl-2-piperidineacetic acid
1-Benzyl-2-piperidineacetic acid
Formel: C₁₄H₁₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12869983
CAS-Nummer: 1184689-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR