Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate

Lieferant: Apollo Scientific

OR470078-5G
APOSOR470078-5GEA 1010 EUR
APOSOR470078-5G
Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate
Methyl-5-bromo-2-(methylsulphonyl)-4-pyrimidinecarboxylate
Formel: C₇H₇BrN₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD11100780
CAS-Nummer: 1060795-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR