N-(4-Pyridinylmethyl)cyclohexanamine

Lieferant: Apollo Scientific

OR470081-5G
APOSOR470081-5GEA 0 EUR
APOSOR470081-5G
N-(4-Pyridinylmethyl)cyclohexanamine
N-(4-Pyridinylmethyl)cyclohexanamine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00980823
CAS-Nummer: 128802-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR