[4-(2-Hydroxyethyl)piperazin-1-yl]acetic acid
Lieferant: Apollo Scientific
OR470082-5G
APOSOR470082-5GEA
322
EUR
APOSOR470082-5G
[4-(2-Hydroxyethyl)piperazin-1-yl]acetic acid
[4-(2-Hydroxyethyl)piperazin-1-yl]acetic acid
Formel:
C₈H₁₆N₂O₃ Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03444540 CAS-Nummer: 124335-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...