3-Aminophthalic acid hydrochloride

Lieferant: Apollo Scientific

OR470092-25G
APOSOR470092-25GEA 97.3 EUR
APOSOR470092-25G
3-Aminophthalic acid hydrochloride
3-Aminophthalic acid hydrochloride
Formel: C₈H₈ClNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00017644
CAS-Nummer: 6946-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR