3-(2-Methoxyphenoxy)propylamine

Lieferant: Apollo Scientific

OR470093-1G OR470093-5G
APOSOR470093-1GEA 214 EUR
APOSOR470093-1G APOSOR470093-5G
3-(2-Methoxyphenoxy)propylamine
3-(2-Methoxyphenoxy)propylamine
Formel: C₁₀H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05053657
CAS-Nummer: 3245-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR