[4-(Methylthio)phenoxy]acetic acid

Lieferant: Apollo Scientific

OR470100-1G OR470100-5G
APOSOR470100-1GEA 194 EUR
APOSOR470100-1G APOSOR470100-5G
[4-(Methylthio)phenoxy]acetic acid
[4-(Methylthio)phenoxy]acetic acid
Formel: C₉H₁₀O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00233485
CAS-Nummer: 15267-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR