N-[4-(5-Isobutyramido-1H-benzoimidazol-2-yl)phenyl]benzamide
Lieferant: Apollo Scientific
OR470111-100MG
APOSOR470111-100MGEA
397
EUR
APOSOR470111-100MG
N-[4-(5-Isobutyramido-1H-benzoimidazol-2-yl)phenyl]benzamide
N-[4-(5-Isobutyramido-1H-benzoimidazol-2-yl)phenyl]benzamide
Formel:
C₂₄H₂₂N₄O₂ Molecular Weight: 398,457 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375239 CAS-Nummer: 1261268-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...