5-[(1-Indolyl)methyl]-3-methylisoxazole
Lieferant: Apollo Scientific
OR470113-1G
APOSOR470113-1GEA
601
EUR
APOSOR470113-1G
5-[(1-Indolyl)methyl]-3-methylisoxazole
5-[(1-Indolyl)methyl]-3-methylisoxazole
Formel:
C₁₃H₁₂N₂O Molecular Weight: 212,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375243 CAS-Nummer: 528593-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...