N-[4-(5-Aminobenzimidazol-2-yl)phenyl]benzamide

Lieferant: Apollo Scientific

OR470115-1G
APOSOR470115-1GEA 397 EUR
APOSOR470115-1G
N-[4-(5-Aminobenzimidazol-2-yl)phenyl]benzamide
N-[4-(5-Aminobenzimidazol-2-yl)phenyl]benzamide
Formel: C₂₀H₁₆N₄O
Molecular Weight: 328,367 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375244
CAS-Nummer: 1027155-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR