4-[2-(Hydroxymethyl)-1H-benzoimidazol-1-yl]butyl Pivalate
Lieferant: Apollo Scientific
OR470116-1G
APOSOR470116-1GEA
397
EUR
APOSOR470116-1G
4-[2-(Hydroxymethyl)-1H-benzoimidazol-1-yl]butyl Pivalate
4-[2-(Hydroxymethyl)-1H-benzoimidazol-1-yl]butyl Pivalate
Formel:
C₁₇H₂₄N₂O₃ Molecular Weight: 304.384 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375245 CAS-Nummer: 797032-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...