(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine

Lieferant: Apollo Scientific

OR470117-1G
APOSOR470117-1GEA 1020 EUR
APOSOR470117-1G
(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
Formel: C₁₈H₃₂N₄S
Molecular Weight: 336,538 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18911728
CAS-Nummer: 1393886-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR