Methyl 2-Bromo-5-sulphamoylbenzoate
Lieferant: Apollo Scientific
OR470123-1G
OR470123-5G
APOSOR470123-1GEA
397
EUR
APOSOR470123-1G
APOSOR470123-5G
Methyl 2-Bromo-5-sulphamoylbenzoate
Methyl 2-Bromo-5-sulphamoylbenzoate
Formel:
C₈H₈BrNO₄S Molecular Weight: 294.122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11057846 CAS-Nummer: 924867-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...