1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid

Lieferant: Apollo Scientific

Synonyme: 1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride

OR470126-25G
APOSOR470126-25GEA 114 EUR
APOSOR470126-25G
1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid
1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid
Formel: C₂₁H₂₅ClN₂
Molecular Weight: 340,89 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684541
CAS-Nummer: 141556-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR