2-(Boc-amino)ethoxylbenzonitrile

Lieferant: Apollo Scientific

OR470128-5G
APOSOR470128-5GEA 591 EUR
APOSOR470128-5G
2-(Boc-amino)ethoxylbenzonitrile
2-(Boc-amino)ethoxylbenzonitrile
Formel: C₁₄H₁₈N₂O₃
Molecular Weight: 262.304 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18070983
CAS-Nummer: 263409-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR