Ethyl 2-(2,4-Dinitrophenyl)-2-methylpropanoate

Lieferant: Apollo Scientific

OR470130-5G
APOSOR470130-5GEA 794 EUR
APOSOR470130-5G
Ethyl 2-(2,4-Dinitrophenyl)-2-methylpropanoate
Ethyl 2-(2,4-Dinitrophenyl)-2-methylpropanoate
Formel: C₁₂H₁₄N₂O₆
Molecular Weight: 282,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18070984
CAS-Nummer: 1256633-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR