3-(Boc-aminoethyloxy)benzonitrile

Lieferant: Apollo Scientific

OR470133-5G
APOSOR470133-5GEA 591 EUR
APOSOR470133-5G
3-(Boc-aminoethyloxy)benzonitrile
3-(Boc-aminoethyloxy)benzonitrile
Formel: C₁₄H₁₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18070988
CAS-Nummer: 252263-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR