1-(Methoxymethyl)-3-nitrobenzene

Lieferant: Apollo Scientific

OR470134-25G OR470134-5G OR470134-10G
APOSOR470134-25GEA 1060 EUR
APOSOR470134-25G APOSOR470134-5G APOSOR470134-10G
1-(Methoxymethyl)-3-nitrobenzene
1-(Methoxymethyl)-3-nitrobenzene
Formel: C₈H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18070994
CAS-Nummer: 1515-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR