4,5-Isoxazoledicarbonitrile

Lieferant: Apollo Scientific

OR470135-5G
APOSOR470135-5GEA 306 EUR
APOSOR470135-5G
4,5-Isoxazoledicarbonitrile
4,5-Isoxazoledicarbonitrile
Formel: C₅HN₃O
Molecular Weight: 119,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18070995
CAS-Nummer: 1256633-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR