2-(Methylsulphonamido)phenylacetonitrile

Lieferant: Apollo Scientific

OR470136-5G
APOSOR470136-5GEA 601 EUR
APOSOR470136-5G
2-(Methylsulphonamido)phenylacetonitrile
2-(Methylsulphonamido)phenylacetonitrile
Formel: C₉H₁₀N₂O₂S
Molecular Weight: 210.253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18070996
CAS-Nummer: 36268-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR