Ethyl 2-(Methylsulphonamido)phenylacetate

Lieferant: Apollo Scientific

OR470137-5G
APOSOR470137-5GEA 601 EUR
APOSOR470137-5G
Ethyl 2-(Methylsulphonamido)phenylacetate
Ethyl 2-(Methylsulphonamido)phenylacetate
Formel: C₁₁H₁₅NO₄S
Molecular Weight: 257,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18070997
CAS-Nummer: 36064-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR