2-(Cyclopentyloxy)acetamide
Lieferant: Apollo Scientific
OR470140-5G
APOSOR470140-5GEA
916
EUR
APOSOR470140-5G
2-(Cyclopentyloxy)acetamide
2-(Cyclopentyloxy)acetamide
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16042597 CAS-Nummer: 855929-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...