Ethyl 3-(4-Isobutylphenyl)acrylate
Lieferant: Apollo Scientific
OR470141-5G
APOSOR470141-5GEA
713
EUR
APOSOR470141-5G
Ethyl 3-(4-Isobutylphenyl)acrylate
Ethyl 3-(4-Isobutylphenyl)acrylate
Formel:
C₁₅H₂₀O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071004 CAS-Nummer: 1256636-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...