Diethyl (hydroxyimino)malonate

Lieferant: Apollo Scientific

Synonyme: Diethyl Isonitrosomalonate

OR470144-5G
APOSOR470144-5GEA 496 EUR
APOSOR470144-5G
Diethyl (hydroxyimino)malonate
Diethyl (hydroxyimino)malonate
Formel: C₇H₁₁NO₅
Molecular Weight: 189,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019949
CAS-Nummer: 6829-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR