Sodium 3-Cyanopropane-1-sulphonate

Lieferant: Apollo Scientific

OR470147-5G
APOSOR470147-5GEA 1010 EUR
APOSOR470147-5G
Sodium 3-Cyanopropane-1-sulphonate
Sodium 3-Cyanopropane-1-sulphonate
Formel: C₄H₆NO₃S·Na
Lagertemperatur: Raumtemperatur
MDL: MFCD18071013
CAS-Nummer: 35880-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR