3-Methoxypropane-1-sulphonic acid

Lieferant: Apollo Scientific

OR470148-5G
APOSOR470148-5GEA 1010 EUR
APOSOR470148-5G
3-Methoxypropane-1-sulphonic acid
3-Methoxypropane-1-sulphonic acid
Formel: C₄H₁₀O₄S
Molecular Weight: 154,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071014
CAS-Nummer: 51980-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR