2-Acetamidophenethyl alcohol

Lieferant: Apollo Scientific

OR470149-1G
APOSOR470149-1GEA 550 EUR
APOSOR470149-1G
2-Acetamidophenethyl alcohol
2-Acetamidophenethyl alcohol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01671553
CAS-Nummer: 69258-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR