1-Methylpiperidine-4-carbohydrazide
Lieferant: Apollo Scientific
OR470150-5G
APOSOR470150-5GEA
997
EUR
APOSOR470150-5G
1-Methylpiperidine-4-carbohydrazide
1-Methylpiperidine-4-carbohydrazide
Formel:
C₇H₁₅N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188558 CAS-Nummer: 176178-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...