Methyl 2-Acetamido-2-phenylacetate

Lieferant: Apollo Scientific

OR470152-5G
APOSOR470152-5GEA 794 EUR
APOSOR470152-5G
Methyl 2-Acetamido-2-phenylacetate
Methyl 2-Acetamido-2-phenylacetate
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16691219
CAS-Nummer: 36061-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR