1-Iodo-4-(methylsulphonyl)benzene

Lieferant: Apollo Scientific

OR470154-1G
APOSOR470154-1GEA 214 EUR
APOSOR470154-1G
1-Iodo-4-(methylsulphonyl)benzene
1-Iodo-4-(methylsulphonyl)benzene
Formel: C₇H₇IO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00955957
CAS-Nummer: 64984-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR