Methyl 4-(2-Ethoxy-2-oxoethylamino)butanoate

Lieferant: Apollo Scientific

OR470157-5G
APOSOR470157-5GEA 601 EUR
APOSOR470157-5G
Methyl 4-(2-Ethoxy-2-oxoethylamino)butanoate
Methyl 4-(2-Ethoxy-2-oxoethylamino)butanoate
Formel: C₉H₁₇NO₄
Molecular Weight: 203,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12148722
CAS-Nummer: 1183267-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR