2-[2-(Boc-amino)ethoxy]benzamide

Lieferant: Apollo Scientific

OR470158-1G OR470158-5G
APOSOR470158-1GEA 306 EUR
APOSOR470158-1G APOSOR470158-5G
2-[2-(Boc-amino)ethoxy]benzamide
2-[2-(Boc-amino)ethoxy]benzamide
Formel: C₁₄H₂₀N₂O₄
Molecular Weight: 280.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071034
CAS-Nummer: 1256633-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR