(R)-2-(N-Cbz-3-pyrrolidinyl)-2-propanol

Lieferant: Apollo Scientific

OR470160-1G
APOSOR470160-1GEA 794 EUR
APOSOR470160-1G
(R)-2-(N-Cbz-3-pyrrolidinyl)-2-propanol
(R)-2-(N-Cbz-3-pyrrolidinyl)-2-propanol
Formel: C₁₅H₂₁NO₃
Molecular Weight: 263.332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071038
CAS-Nummer: 1417789-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR