(R)-1-Cbz-3-pyrrolidinecarboxamide

Lieferant: Apollo Scientific

OR470161-1G
APOSOR470161-1GEA 561 EUR
APOSOR470161-1G
(R)-1-Cbz-3-pyrrolidinecarboxamide
(R)-1-Cbz-3-pyrrolidinecarboxamide
Formel: C₁₃H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09878793
CAS-Nummer: 1217835-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR