Methyl 3-chloro-4-(4-methylpiperazino)benzoate

Lieferant: Apollo Scientific

OR470163-5G
APOSOR470163-5GEA 601 EUR
APOSOR470163-5G
Methyl 3-chloro-4-(4-methylpiperazino)benzoate
Methyl 3-chloro-4-(4-methylpiperazino)benzoate
Formel: C₁₃H₁₇ClN₂O₂
Molecular Weight: 268,739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071042
CAS-Nummer: 1256633-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR