Ethyl 3-(Methylsulphonyl)pyridine-2-carboxylate

Lieferant: Apollo Scientific

OR470164-5G
APOSOR470164-5GEA 1330 EUR
APOSOR470164-5G
Ethyl 3-(Methylsulphonyl)pyridine-2-carboxylate
Ethyl 3-(Methylsulphonyl)pyridine-2-carboxylate
Formel: C₉H₁₁NO₄S
Molecular Weight: 229,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071045
CAS-Nummer: 1256633-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR