Ethyl 5,8-Dichloroimidazo[1,5-a]pyrazine-1-carboxylate

Lieferant: Apollo Scientific

OR470166-1G
APOSOR470166-1GEA 1330 EUR
APOSOR470166-1G
Ethyl 5,8-Dichloroimidazo[1,5-a]pyrazine-1-carboxylate
Ethyl 5,8-Dichloroimidazo[1,5-a]pyrazine-1-carboxylate
Formel: C₉H₇Cl₂N₃O₂
Molecular Weight: 260,077 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071050
CAS-Nummer: 1256633-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR