4,6-Dihydroxynicotinamide

Lieferant: Apollo Scientific

OR470170-1G OR470170-5G
APOSOR470170-1GEA 199 EUR
APOSOR470170-1G APOSOR470170-5G
4,6-Dihydroxynicotinamide
4,6-Dihydroxynicotinamide
Formel: C₆H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18651680
CAS-Nummer: 5466-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR