6-Phenoxynicotinonitrile

Lieferant: Apollo Scientific

OR470172-1G OR470172-5G
APOSOR470172-1GEA 92.8 EUR
APOSOR470172-1G APOSOR470172-5G
6-Phenoxynicotinonitrile
6-Phenoxynicotinonitrile
Formel: C₁₂H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00205454
CAS-Nummer: 99902-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR