2-Brom-5-sulfamoylbenzoesäure
Lieferant: Apollo Scientific
OR470176-1G
APOSOR470176-1GEA
397
EUR
APOSOR470176-1G
2-Brom-5-sulfamoylbenzoesäure
2-Brom-5-sulfamoylbenzoesäure
Formel:
C₇H₆BrNO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD06355953 CAS-Nummer: 22361-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...