Methyl N-Boc-2-bromo-5-sulphamoylbenzoate

Lieferant: Apollo Scientific

OR470177-1G
APOSOR470177-1GEA 397 EUR
APOSOR470177-1G
Methyl N-Boc-2-bromo-5-sulphamoylbenzoate
Methyl N-Boc-2-bromo-5-sulphamoylbenzoate
Formel: C₁₃H₁₆BrNO₆S
Molecular Weight: 394,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651694
CAS-Nummer: 1272756-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR