1-Isopropyl-2-oxopyrrolidine-4-carboxamidine hydrochloride
Lieferant: Apollo Scientific
OR470181-1G
APOSOR470181-1GEA
601
EUR
APOSOR470181-1G
1-Isopropyl-2-oxopyrrolidine-4-carboxamidine hydrochloride
1-Isopropyl-2-oxopyrrolidine-4-carboxamidine hydrochloride
Formel:
C₈H₁₅N₃O·ClH Molecular Weight: 205,685 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651697 CAS-Nummer: 1272756-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...