1-(2-Methoxyethyl)-2-oxopyrrolidine-4-carboxamidine hydrochloride

Lieferant: Apollo Scientific

OR470182-1G
APOSOR470182-1GEA 601 EUR
APOSOR470182-1G
1-(2-Methoxyethyl)-2-oxopyrrolidine-4-carboxamidine hydrochloride
1-(2-Methoxyethyl)-2-oxopyrrolidine-4-carboxamidine hydrochloride
Formel: C₈H₁₅N₃O₂·ClH
Molecular Weight: 221,685 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651698
CAS-Nummer: 1272756-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR