2-(5-Oxazolyl)benzylamine

Lieferant: Apollo Scientific

OR470183-1G OR470183-5G
APOSOR470183-1GEA 397 EUR
APOSOR470183-1G APOSOR470183-5G
2-(5-Oxazolyl)benzylamine
2-(5-Oxazolyl)benzylamine
Formel: C₁₀H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18375280
CAS-Nummer: 1261268-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR