4-(4-Nitrophenyl)thiazole-2-carboxylic acid

Lieferant: Apollo Scientific

OR470184-1G
APOSOR470184-1GEA 824 EUR
APOSOR470184-1G
4-(4-Nitrophenyl)thiazole-2-carboxylic acid
4-(4-Nitrophenyl)thiazole-2-carboxylic acid
Formel: C₁₀H₆N₂O₄S
Molecular Weight: 250,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375281
CAS-Nummer: 4415-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR